Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Zytron 200 Totally Encapsulating Suits

$310.00 - $695.00

Specifications

Color Gray
Material Zytron™ 200
For Use With (Application) Ideally used with wider range of chemicals
Resistance Chemical
View More Specs

Products 20
Catalog Number Mfr. No. Description Seam Type Size Price Quantity & Availability  
Catalog Number Mfr. No. Description Seam Type Size Price Quantity & Availability  
19-171-144A
View Documents
Kappler™
Z2B371GYSMMD
Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped Medium
Case of 6 for $310.00
Add to cart
 
19-171-144B
View Documents
Kappler™
Z2B371GYLGXL
Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped Large
Case of 6 for $310.00
Add to cart
 
19-171-144D
View Documents
Kappler™
Z2B371GY2X3X
Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped 2X-Large
Case of 6 for $341.00
Add to cart
 
19-171-144C
View Documents
Kappler™
Z2B371GYSLGXL
Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped X-Large
Case of 6 for $310.00
Add to cart
 
19-171-145A
View Documents
Kappler™
Z2H371GYSMMD
Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped Medium
Case of 6 for $612.00
Add to cart
 
19-171-145B
View Documents
Kappler™
Z2H371GYLGXL
Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped Large
Case of 6 for $612.00
Add to cart
 
19-171-144E
View Documents
Kappler™
Z2B3712X3X
Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped 3X-Large
Case of 6 for $341.00
Add to cart
 
19-171-145C
View Documents
Kappler™
Z2H371XL
Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped X-Large
Case of 6 for $612.00
Add to cart
 
19-171-145D
View Documents
Kappler™
Z2H371GY2X3X
Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped 2X-Large
Case of 6 for $673.00
Add to cart
 
19-171-145E
View Documents
Kappler™
Z2H371GYS2X3X
Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped 3X-Large
Case of 6 for $673.00
Add to cart
 
19-171-146B
View Documents
Kappler™
Z2B571GYLGXL
Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped Large
Case of 6 for $351.00
Add to cart
 
19-171-146C
View Documents
Kappler™
Z2B571GYSLGXL
Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped X-Large
Case of 6 for $351.00
Add to cart
 
19-171-146D
View Documents
Kappler™
Z2B571GY2X3X
Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped 2X-Large
Case of 6 for $386.00
Add to cart
 
19-171-146A
View Documents
Kappler™
Z2B571GYSMMD
Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped Medium
Case of 6 for $351.00
Add to cart
 
19-171-147A
View Documents
Kappler™
Z2H571GYSMMD
Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped Medium
Case of 6 for $632.00
Add to cart
 
19-171-147B
View Documents
Kappler™
Z2H571GYLGXL
Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped Large
Case of 6 for $632.00
Add to cart
 
19-171-146E
View Documents
Kappler™
Z2B5713X
Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped 3X-Large
Case of 6 for $386.00
Add to cart
 
19-171-147C
View Documents
Kappler™
Z2H571XL
Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped X-Large
Case of 6 for $632.00
Add to cart
 
19-171-147D
View Documents
Kappler™
Z2H571GY2X3X
Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped 2X-Large
Case of 6 for $695.00
Add to cart
 
19-171-147E
View Documents
Kappler™
Z2H5713X
Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped, Taped 3X-Large
Case of 6 for $695.00
Add to cart
 
Description

Description

Specifications

Specifications

Gray
Zytron™ 200
Ideally used with wider range of chemicals
Chemical
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit