Alfa Aesar™ Karl Fischer Coulometric AG, anolyte solution, for cells with or without diaphragm

Manufacturer: Alfa Aesar™ 47106AP

Quantity 500mL

 View more versions of this product

Catalog No. AA47106AP

  Check Availability
Add to Cart

    Chemical Identifiers

    UN Number UN3286
    Chemical Name or Material Karl Fischer Coulometric AG, anolyte solution, for cells with or without diaphragm

    Specifications

    Quantity 500mL
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H225-H301-H311-H331-H370-H314
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.
    Causes severe skin burns and eye damage.
    UN Number UN3286
    DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
    Density 0.930g/mL
    Boiling Point 63°C
    Color Colorless to pale yellow
    Health Hazard 1 Danger
    Chemical Name or Material Karl Fischer Coulometric AG, anolyte solution, for cells with or without diaphragm

    GHS H Statement
    H225-H301-H311-H331-H370-H314
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit