Alfa Aesar™ Karl Fischer Coulometric CG, catholyte solution

Manufacturer: Alfa Aesar™ 47117PDL

Quantity 110mL

 View more versions of this product

Catalog No. AA47117PDL

  Check Availability
Add to Cart

    Chemical Identifiers

    UN Number UN3267
    Chemical Name or Material Karl Fischer Coulometric CG, Catholyte solution

    Specifications

    Quantity 110mL
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3267
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H302-H227
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Combustible liquid.
    Physical Form Liquid
    Density 1.1g/mL
    Sensitivity Moisture Sensitive
    Boiling Point 202°C
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    Color Colorless to pale yellow
    Chemical Name or Material Karl Fischer Coulometric CG, Catholyte solution

    GHS H Statement
    H314-H302-H227
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit