Alfa Aesar™ Karl Fischer Methanol, solvent/working medium for volumetric one-component titration

Manufacturer: Alfa Aesar™ 47165Y0

Quantity 6 x 1L

 View more versions of this product

Catalog No. AA47165Y0

  Check Availability
Add to Cart

    Chemical Identifiers

    UN Number UN1230
    Chemical Name or Material Karl Fischer Methanol, solvent/working medium for volumetric one-component titration

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 65°C
    Physical Form Liquid
    Color Colorless to pale green
    Quantity 6 x 1L
    UN Number UN1230
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H311-H331-H370
    Highly flammable liquid and vapor.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.
    Density 0.910g/mL
    DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
    Chemical Name or Material Karl Fischer Methanol, solvent/working medium for volumetric one-component titration

    GHS H Statement
    H225-H311-H331-H370
    Highly flammable liquid and vapor.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.

    GHS P Statement
    P210-P260-P303+P361+P353-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit