Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Karl Fischer Reagent Diluent, Fisher Chemical
For SK3 stabilized Karl Fischer Reagent
$463.00 - $1117.00

Specifications
Chemical Name or Material | Karl Fischer Reagent Diluent |
---|---|
CAS | 109-86-4, 110-86-1 |
MDL Number | MFCD00081734 |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
SK5-1
|
1L | Glass Bottle |
Each for $463.00
Case of 6 EA for $2,222.01
Order in Bulk
Request bulk or custom formats |
|
SK5-4
|
4L | Glass Bottle |
Each for $1,117.00
Case of 4 EA for $3,572.48
Order in Bulk
Request bulk or custom formats |
|
Description
EcoSafPak is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Karl Fischer Reagent Diluent | |
109-86-4, 110-86-1 | |
MFCD00081734 |
Safety and Handling

DOTInformation : DOT Class 3, : Flammable Liquid
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title