Learn More
Karl Fischer Reagent Diluent, Fisher Chemical™
For SK3 stabilized Karl Fischer Reagent
 View more versions of this product
Catalog No. SK5-1
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
Specifications
Specifications
109-86-4,110-86-1
Yellow
Pass Test
Glass Bottle
1L
Karl Fischer Reagent Diluent
0.93g/cm³
MFCD00081734
Liquid
Safety and Handling
Safety and Handling
DOTInformation : DOT Class 3, : Flammable Liquid