Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Karl Fischer Reagent Diluent, Fisher Chemical™
For SK3 stabilized Karl Fischer Reagent
Specifications
109-86-4,110-86-1 | |
Yellow | |
Pass Test | |
Glass Bottle | |
4L |
Karl Fischer Reagent Diluent | |
0.93g/cm³ | |
MFCD00081734 | |
Liquid |
Safety and Handling

DOTInformation : DOT Class 3, : Flammable Liquid
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title