Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Karl Fischer reagent, stabilized, Thermo Scientific Chemicals
Specifications
Density | 0.93g/mL |
---|---|
Chemical Name or Material | Karl Fischer reagent |
CAS | 7553-56-2 |
Dilution Test | Pass Test |
Equivalence | 6 EQUAL min. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AC611130010
|
Thermo Scientific Chemicals
611130010 |
1 L | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | |||||
AC611130040
|
Thermo Scientific Chemicals
611130040 |
4 L | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
0.93g/mL | |
7553-56-2 | |
6 EQUAL min. | |
MFCD00011355 MFCD00164163 | |
II | |
253.81 | |
CHEBI:17606 |
Karl Fischer reagent | |
Pass Test | |
I2 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
diiodine | |
807 |
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title