Alfa Aesar™ Karl Fischer Solvent CS, solvent for volumetric titration, for oils

Manufacturer: Alfa Aesar™ 47217Y0

Quantity 6 x 1L

 View more versions of this product

Catalog No. AA47217Y0

  Check Availability
Add to Cart

    Chemical Identifiers

    UN Number UN3286
    Chemical Name or Material Karl Fischer Solvent CS, solvent for volumetric titration, for oils

    Specifications

    Quantity 6 x 1L
    Health Hazard 2 GHS H Statement
    H225-H311-H331-H370-H351-H373-H314-H302
    Highly flammable liquid and vapor.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.
    Suspected of causing cancer.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    UN Number UN3286
    Health Hazard 1 Danger
    Density 1.040g/mL
    Boiling Point 59°C
    DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
    Color Colorless to pale yellow
    Chemical Name or Material Karl Fischer Solvent CS, solvent for volumetric titration, for oils

    GHS H Statement
    H225-H311-H331-H370-H351-H373-H314-H302
    Highly flammable liquid and vapor.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.
    Suspected of causing cancer.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit