Learn More
Karl Fischer Solvent KT, solvent for volumetric one- and two-component titration, for aldehydes and ketones, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 47219K2
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Colorless to Yellow | |
1 L | |
solvent for volumetric one- and two-component titration, for aldehydes and ketones | |
Liquid |
Karl Fischer Solvent KT | |
0.940g/mL | |
120°C | |
UN2924 |
Safety and Handling
GHS H Statement
H314-H226-H336
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : TSCA
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.