Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Volumetric Reagent, MilliporeSigma™

Specifications

Chemical Name or Material Karl Fischer Volumetric Reagent
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
MKX00106
View Documents
MilliporeSigma
KX00106
1L Poly-coated Amber Glass
Each for $353.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Reagent for Volumetric Karl Fischer Titrations.
  • For titrimetric moisture determination. Titer ≈ 5mg H2O/mL.
  • Contains pyridine and 2-methoxyethanol.
Specifications

Specifications

Karl Fischer Volumetric Reagent
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit