Learn More
Dynalon™ Kartell Bucket with Graduations and Spout Products for Science Education
For measuring and transporting liquids or solids. Dynalon™ Kartell Bucket with Graduations and Spout are chemical resistant especially at room temperature.
$18.75 - $35.75
Specifications
Material LDPE
Graduated Yes
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Height (English) Price Quantity & Availability  
S37255 Products for Science Education
View Documents
Dynalon™
2131850009
2.37 gal. 9.84 in. 11.02 in.
Each for $18.75
Add to cart
S37256 Products for Science Education
View Documents
Dynalon™
2131850012
3.17 gal. 11.02 in. 11.81 in.
Each for $24.50
Add to cart
S37257 Products for Science Education
View Documents
Dynalon™
2131850017
4.49 gal. 12.2 in. 13.38 in.
Each for $35.75
Add to cart
Description
Description
  • Features steel bale handle with comfortable grip
  • Made of LDPE
  • Graduated internally in liters
  • Use up to 80˚C or well below freezing
Specifications
Specifications
LDPE
Yes