Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Kasugamycin, 50mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
25mL
50mL
19408-46-9
C14H26ClN3O9
415.824
MFCD00058459
ZDRBJJNXJOSCLR-YZKQBBCCSA-N
66765861
2-amino-2-[(2R,3S,5S,6R)-5-amino-2-methyl-6-[(2R,3S,5S,6S)-2,3,4,5,6-pentahydroxycyclohexyl]oxyoxan-3-yl]iminoacetic acid;hydron;chloride
[H+].CC1C(CC(C(O1)OC2C(C(C(C(C2O)O)O)O)O)N)N=C(C(=O)O)N.[Cl-]
19408-46-9
C14H26ClN3O9
415.824
MFCD00058459
ZDRBJJNXJOSCLR-YZKQBBCCSA-N
66765861
2-amino-2-[(2R,3S,5S,6R)-5-amino-2-methyl-6-[(2R,3S,5S,6S)-2,3,4,5,6-pentahydroxycyclohexyl]oxyoxan-3-yl]iminoacetic acid;hydron;chloride
[H+].CC1C(CC(C(O1)OC2C(C(C(C(C2O)O)O)O)O)N)N=C(C(=O)O)N.[Cl-]
Odor Odorless
Physical Form Liquid
Quantity 50mL
Chemical Name or Material Kasugamycin, 50mg/mL in distilled water, sterile-filtered

RTECSNumber : NM7521800

storageNote1 : -30° to -10°C