Learn More
KAT2A/GCN5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149831AP
Catalog No. 50-172-9885
Add to cart
Description
Description
SAGA (Spt-Ada-Gen5 histone acetyltransferase complex) is a histoneacetylase complex which has Gcn5p as catalyst subunit and functions overlapping with the fundamental transcription factor TFIID which has Taf1p as catalyst subunit. However, SAGA and TFIID have different allotment and each accomplishes the important role in the transcription for Housekeeping gene group and Stress Responding gene group. Also, SAGA is a gigantic protein complex which is composed of Ada protein group (5 kinds), TBP related protein group (4 kinds), TAF protein group (5 kinds that also pertain to TFIID), and other protein groups (>6 kinds). As its molecular function, there is the chemical modification of histone, or the recruitment by direct interaction of transcription regulating factor on DNA, or the control of transcription starting reaction by TBP. Gcn5p that show histoneacetylase activity is one kind of the above-mentioned Ada protein group, and in the case of budding yeast it is composed of 439 amino acid residues.
Specifications
Specifications
KAT2A/GCN5
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q92830, Q9JHD2
GCN5, GCN5L2, hGCN5, Histone acetyltransferase GCN5, HsGCN5, K(lysine) acetyltransferase 2A, KAT2A, Lysine acetyltransferase 2A, PCAF b, STAF97
Rabbit
IgG
150 μL
-20°C
Primary
14534, 2648
Western Blot
Unconjugated
KAT2A
Liquid
Kat2a
KAT2A/GCN5 Fusion Protein Ag6940
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse