Learn More
KAT3B/p300 Mouse anti-Human, Clone: 1H11G2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259233
Catalog No. NBP259233
Add to cart
Description
Description
KAT3B/p300 Monoclonal antibody specifically detects KAT3B/p300 in Human samples. It is validated for Chromatin Immunoprecipitation.
Specifications
Specifications
KAT3B/p300
1H11G2
Chromatin Immunoprecipitation 5 ug
Purified
EP300
The exact immunogen is propietary information.
Protein A purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ChIP assay
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2
Mouse
IgG3
50 ug
Adaptive Immunity, Alzheimers Research, Amino Acids Drugs and other small molecules, Angiogenesis, Apoptosis, Asthma, Astrocyte Markers, AU1 and AU5 Epitope Tags, Autophagy, Base Excision Repair, Beta Galactosidase Epitope Tags, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Vision, Wnt Signaling Pathway
Primary
2033