Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

katanin-p80 Mouse anti-Human, PE/Cy5.5, Clone: OTI5C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271578PECY55

Catalog No. N271578PEC5


Add to cart

Description

Description

katanin-p80 Monoclonal antibody specifically detects katanin-p80 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

katanin-p80
OTI5C4
Flow Cytometry
Affinity Purified, Conjugated
KATNB1
Full length recombinant protein of human katanin-p80 (NP_005877) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
10300.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
KAT, katanin (80 kDa), katanin p80 (WD repeat containing) subunit B 1, katanin p80 (WD40-containing) subunit B 1, Katanin p80 subunit B1, katanin p80 WD40-containing subunit B1, p80 katanin
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit