Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

katanin-p80 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen katanin-p80
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB124932
View Documents
Novus Biologicals
NBP31001525UL
25 μg
Each for $106.19
Add to cart
 
NB124931
View Documents
Novus Biologicals
NBP310015100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

katanin-p80 Polyclonal antibody specifically detects katanin-p80 in Human samples. It is validated for Western Blot
Specifications

Specifications

katanin-p80
Western Blot
Purified
RUO
Human
KAT, katanin (80 kDa), katanin p80 (WD repeat containing) subunit B 1, katanin p80 (WD40-containing) subunit B 1, Katanin p80 subunit B1, katanin p80 WD40-containing subunit B1, p80 katanin
The immunogen is a synthetic peptide directed towards the middle region of human katanin-p80 (NP_005877.2). Peptide sequence RVKQNSESERRSPSSEDDRDERESRAEIQNAEDYNEIFQPKNSISRTPPR
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Stem Cell Markers
PBS buffer, 2% sucrose
10300
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit