Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KATNAL1 Mouse anti-Human, Clone: OTI6A4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525453

Catalog No. PIMA525453


Add to cart

Description

Description

Regulates microtubule dynamics in Sertoli cells, a process that is essential for spermiogenesis and male fertility. Severs microtubules in an ATP-dependent manner, promoting rapid reorganization of cellular microtubule arrays
Specifications

Specifications

KATNAL1
Monoclonal
0.45 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BW62
katanin p60 ATPase-containing subunit A-like 1, p60 katanin-like 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84056
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI6A4
Unconjugated
KATNAL1
Liquid
KATNAL1
Full length human recombit protein of KATNAL1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit