Learn More
KATNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149691AP
Catalog No. 50-172-9874
Add to cart
Description
Description
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. Katanin is a member of the AAA family of ATPases.
Specifications
Specifications
KATNB1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BG40, Q9BVA0
KAT, Katanin p80 subunit B1, KATNB1, p80 katanin
Rabbit
IgG
150 μL
-20°C
Primary
10300, 291852, 74187
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
KATNB1
Liquid
Katnb1
KATNB1 Fusion Protein Ag6836
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat