Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Katanin p80 Monoclonal Antibody (OTI5C4), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525743
Description
Katanin p80 Monoclonal antibody specifically detects Katanin p80 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Katanin p80 | |
Monoclonal | |
0.64 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BVA0 | |
Katnb1 | |
Full length human recombinant protein of KATNB1 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI5C4 | |
Unconjugated | |
Katnb1 | |
2410003J24Rik; KAT; katanin (80 kDa); katanin p80 (WD repeat containing) subunit B 1; katanin p80 (WD40-containing) subunit B 1; katanin p80 subunit B1; Katanin p80 WD40 repeat-containing subunit B1; katanin p80 WD40-containing subunit B1; katanin regulatory subunit B1; KATNB1; LIS6; p80 katanin | |
Mouse | |
Affinity Chromatography | |
RUO | |
10300 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction