Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kazrin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549095

Catalog No. PA549095

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.
Specifications

Specifications

Kazrin
Polyclonal
PBS with 0.09% sodium azide
Q674X7
Kazrin, KAZN, KAZ, KIAA1026
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23254
Western Blot
Unconjugated
KAZN
Liquid
KAZN
KLH conjugated synthetic peptide between 711-739 amino acids from the C-terminal region of human KAZ.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit