Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kazrin Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551202

Catalog No. PA551202


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total KAZ / KAZN protein.

Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kazrin
Polyclonal
Unconjugated
KAZN
2310007B04Rik; 9030409G11Rik; AI256607; AI666764; AW061099; HRIHFB2003; KAZ; KAZN; Kazrin; kazrin, periplakin interacting protein; KIAA1026
Rabbit
Antigen Affinity Chromatography
RUO
23254
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.4 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q674X7
KAZN
Fusion protein of human KAZN.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.