Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kazrin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556926
Description
Immunogen sequence: RLQEEVHLLR QMKEMLAKDL EESQGGKSSE VLSATELRVQ LAQKEQELAR AKEALQAMKA DRKRLKGEKT DLVSQMQQLY ATLE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.
Specifications
Kazrin | |
Polyclonal | |
Unconjugated | |
KAZN | |
2310007B04Rik; 9030409G11Rik; AI256607; AI666764; AW061099; HRIHFB2003; KAZ; KAZN; Kazrin; kazrin, periplakin interacting protein; KIAA1026 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23254 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q674X7 | |
KAZN | |
Recombinant protein corresponding to Human Kazrin. Recombinant protein control fragment (Product #RP-94541). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction