Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KB SRC 4, Tocris Bioscience™

Catalog No. p-4816566
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective c-Src inhibitor

Potent and selective c-Src inhibitor (Ki = 44 nM); does not inhibit c-Abl at concentrations up to 125 μM. Selective between Src family members (Kd values are 86, 160, 240, 720, 3200, 4400, and >40,000 nM for c-Src, Lck, Fgr, Yes, Lyn, Hck and Fyn respectively). Significantly inhibits cell growth in 4T1 mammary carcinoma tumor cells.

Chemical Identifiers

CAS 1380088-03-8
Molecular Formula C32H23ClN8
Molecular Weight (g/mol) 555.042
InChI Key UHIZYQVRKSWIFO-UHFFFAOYSA-N
Synonym kb src 4, 1-3-1-1,1'-biphenyl-3-ylmethyl-1h-1,2,3-triazol-5-yl phenyl-3-4-chlorophenyl-1h-pyrazolo 3,4-d pyrimidin-4-amine, 1-3-1-1,1-biphenyl-3-ylmethyl-1h-1,2,3-triazol-5-yl phenyl-3-4-chlorophenyl-1h-pyrazolo 3,4-d pyrimidin-4-amine
PubChem CID 73349077
IUPAC Name 3-(4-chlorophenyl)-1-[3-[3-[(3-phenylphenyl)methyl]triazol-4-yl]phenyl]pyrazolo[3,4-d]pyrimidin-4-amine
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)CN3C(=CN=N3)C4=CC(=CC=C4)N5C6=C(C(=N5)C7=CC=C(C=C7)Cl)C(=NC=N6)N

Specifications

Quantity 10 mg
Formula Weight 555.03
Percent Purity >98%
Chemical Name or Material KB SRC 4
Recommended Storage Store at +4°C
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.