Learn More
Abnova™ KBTBD6 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00089890T01
Catalog No. 89-949-483
Add to cart
Specifications
Specifications
293T
NM_152903
pCMV-KBTBD6 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KBTBD6 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 76.1kDa
Transient overexpression cell lysate was tested with anti-KBTBD6 antibody (Mfr. No. H00089890-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human