Learn More
Abnova™ KBTBD9 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Manufacturer:  Abnova™ H00114818T03
Catalog No. 89-949-491
Add to cart
Specifications
Specifications
293T
BC015667
pCMV-KBTBD9 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KBTBD9 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 55.2kDa
Transient overexpression cell lysate was tested with anti-KBTBD9 antibody (Mfr. No. H00114818-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human