Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KC 12291 hydrochloride, Tocris Bioscience™

Catalog Number p-4784067
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
181936-98-1
C22H28ClN3O3S
449.994
KIJPRQYBNQKUAU-UHFFFAOYSA-N
kc 12291 hydrochloride, 3,4-dimethoxy-n-methyl-n-3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propyl benzeneethanamine hydrochloride, 3,4-dimethoxy-n-methyl-n-3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propyl benzeneethanaminehydrochloride, n-3,4-dimethoxyphenethyl-n-methyl-3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propan-1-amine hydrochloride, 2-3,4-dimethoxyphenyl ethyl methyl 3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propyl amine hydrochloride, n-2-3,4-dimethoxyphenyl ethyl-n-methyl-3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propan-1-amine-hydrogen chloride 1/1
22902387
N-[2-(3,4-dimethoxyphenyl)ethyl]-N-methyl-3-[(5-phenyl-1,2,4-thiadiazol-3-yl)oxy]propan-1-amine;hydrochloride
CN(CCCOC1=NSC(=N1)C2=CC=CC=C2)CCC3=CC(=C(C=C3)OC)OC.Cl
This item is not returnable. View return policy
181936-98-1
C22H28ClN3O3S
449.994
KIJPRQYBNQKUAU-UHFFFAOYSA-N
kc 12291 hydrochloride, 3,4-dimethoxy-n-methyl-n-3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propyl benzeneethanamine hydrochloride, 3,4-dimethoxy-n-methyl-n-3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propyl benzeneethanaminehydrochloride, n-3,4-dimethoxyphenethyl-n-methyl-3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propan-1-amine hydrochloride, 2-3,4-dimethoxyphenyl ethyl methyl 3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propyl amine hydrochloride, n-2-3,4-dimethoxyphenyl ethyl-n-methyl-3-5-phenyl-1,2,4-thiadiazol-3-yl oxy propan-1-amine-hydrogen chloride 1/1
22902387
N-[2-(3,4-dimethoxyphenyl)ethyl]-N-methyl-3-[(5-phenyl-1,2,4-thiadiazol-3-yl)oxy]propan-1-amine;hydrochloride
CN(CCCOC1=NSC(=N1)C2=CC=CC=C2)CCC3=CC(=C(C=C3)OC)OC.Cl

Orally active atypical Na+ channel blocker; cardioprotective

Orally active atypical voltage-gated sodium channel blocker. Inhibits sustained sodium currents (INaL) and prevents diastolic contracture in isolated atria in vitro (IC50 values are 9.6 and 0.55 - 0.79 μM respectively). Displays anti-ischemic, bradycardic and cardioprotective activity in vivo.
Quantity 10 mg
Formula Weight 449.99
Percent Purity >97%
Chemical Name or Material KC 12291 hydrochloride
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit