Learn More
Invitrogen™ KC/CXCL1 Mouse ELISA Kit
Sandwich ELISA Kit
$600.00 - $4465.00
Specifications
Accession Number | P12850 |
---|---|
Assay Sensitivity | 1 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EMCXCL1
|
Invitrogen™
EMCXCL1 |
96 Tests |
Pack for $600.00
|
|
|||||
EMCXCL1X10
|
Invitrogen™
EMCXCL1X10 |
10 x 96 Tests |
Each for $4,465.00
|
|
|||||
EMCXCL1X5
|
Invitrogen™
EMCXCL1X5 |
5 x 96 Tests |
Each for $2,930.00
|
|
|||||
Description
The Mouse KC/CXCL1 ELISA quantitates Ms KC in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms KC. Principle of the method The Mouse KC solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.Specifications
P12850 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Mouse | |
Colorimetric Microplate Reader | |
Fsp,Gro1,KC,Mgsa,N51,Scyb1,gro | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
1 pg/mL | |
Biotin | |
Chemokine & Receptors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
14825 | |
<12% | |
HRP | |
Immunology | |
Fsp, Gro1, KC, Mgsa, N51, Scyb1, gro | |
1 hr. 20 min. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.