Learn More
Kimberly-Clark Professional™ KC200 Isolation Cover Gowns
Light fluid contact and contact isolation
Manufacturer:  Kimberly-Clark Professional™ 69988
Catalog No. 19-077-546
Add to cart
Description
Description
  • Made of a medium-weight protective 3-layer SMS fabric and are for facility-wide use when risk of exposure to fluid is between low and moderate
  • Gathered back
  • Tie waist
  • Tape-tab neck closure
  • Meets AAMI Level 2 requirements
Certifications
Meets AAMI Level 2 requirements
Specifications
Specifications
Safety
Tape-tab at neck
100/Cs.
Elastic
Yellow