Learn More
Kimberly-Clark Professional™ KC300 Kimguard™ Sterilization Wrap
Two sheets bonded together, making it possible to double wrap in single step
Specifications
Sterility Non-sterile
Sterilization Method Steam and EO
View More Specs
Products 4
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-077-532
View Documents
Kimberly-Clark Professional™
12836
36 x 36 in. 36 x 36 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-077-537
View Documents
Kimberly-Clark Professional™
12872
54 x 72 in. 54 x 72 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-077-542
View Documents
Kimberly-Clark Professional™
12848
48 x 48 in. 48 x 48 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-077-543
View Documents
Kimberly-Clark Professional™
12845
45 x 45 in. 45 x 45 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Two layers of wrap, thermally sealed along the sides
  • Delivers the protection of double-wrapping in a single step, in about half the time it takes to wrap or open packages using sequential double wrapping
  • Quick way to ensure that instrument sterility is uncompromised
  • Microbial barrier protection with patented Powerguard™ technology
  • Made in USA
  • Pre-Vacuum Steam and Ethylene Oxide (EO) Compatible
  • Flame Resistance: Meets NFPA Class 1
  • Blue
Specifications
Specifications
Non-sterile
Steam and EO