Learn More
Kimberly-Clark Professional™ KC400 Kimguard™ Sterilization Wrap
Two sheets bonded together, making it possible to double wrap in single step
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-077-529
|
Kimberly-Clark Professional™
62240 |
40 x 40 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Two layers of wrap, thermally sealed along the sides
- Delivers the protection of double-wrapping in a single step, in about half the time it takes to wrap or open packages using sequential double wrapping
- Quick way to ensure that instrument sterility is uncompromised
- Microbial barrier protection with patented Powerguard™ technology
- Made in USA
- Pre-Vacuum Steam and Ethylene Oxide (EO) Compatible
- Flame Resistance: Meets NFPA Class 1
- Blue