Learn More
Kimberly-Clark Professional™ KC500 Kimguard™ Sterilization Wrap
Two sheets bonded together, making it possible to double wrap in single step
Manufacturer:  Kimberly-Clark Professional™ 62124
 View more versions of this product
Catalog No. 19-077-525
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Two layers of wrap, thermally sealed along the sides
  • Delivers the protection of double-wrapping in a single step, in about half the time it takes to wrap or open packages using sequential double wrapping
  • Quick way to ensure that instrument sterility is uncompromised
  • Microbial barrier protection with patented Powerguard™ technology
  • Made in USA
  • Pre-Vacuum Steam and Ethylene Oxide (EO) Compatible
  • Flame Resistance: Meets NFPA Class 1
  • Blue
Specifications
Specifications
24 x 24 in.