Learn More
Kimberly-Clark Professional™ KC500 Kimguard™ Sterilization Wrap
Two sheets bonded together, making it possible to double wrap in single step
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-077-525
View Documents
Kimberly-Clark Professional™
62124
24 x 24 in. 24 x 24 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-077-540
View Documents
Kimberly-Clark Professional™
62148
48 x 48 in. 48 x 48 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Two layers of wrap, thermally sealed along the sides
  • Delivers the protection of double-wrapping in a single step, in about half the time it takes to wrap or open packages using sequential double wrapping
  • Quick way to ensure that instrument sterility is uncompromised
  • Microbial barrier protection with patented Powerguard™ technology
  • Made in USA
  • Pre-Vacuum Steam and Ethylene Oxide (EO) Compatible
  • Flame Resistance: Meets NFPA Class 1
  • Blue