Learn More
Kimberly-Clark Professional™ KC600 Kimguard™ Sterilization Wrap
Two sheets bonded together, making it possible to double wrap in single step
Manufacturer: Kimberly-Clark Professional™ 62640
Description
- Two layers of wrap, thermally sealed along the sides
- Delivers the protection of double-wrapping in a single step, in about half the time it takes to wrap or open packages using sequential double wrapping
- Quick way to ensure that instrument sterility is uncompromised
- Microbial barrier protection with patented Powerguard™ technology
- Made in USA
- Pre-Vacuum Steam and Ethylene Oxide (EO) Compatible
- Flame Resistance: Meets NFPA Class 1
- Blue
Specifications
Autoclavable | |
40 x 40 in. | |
Non-sterile |
Blue | |
Steam and EO |