Learn More
KCC2/SLC12A5 Mouse anti-Human, Alexa Fluor 700, Clone: 905705, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8369N100UG
Catalog No. FAB8369N100
Add to cart
This item is not returnable. View return policy
Description
Description
KCC2/SLC12A5 Monoclonal antibody specifically detects KCC2/SLC12A5 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
KCC2/SLC12A5
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q9H2X9
SLC12A5
NS0 mouse myeloma cell line transfected with human SLC12A5 Accession # Q9H2X9
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human SLC12A5 in ELISAs and Western blots.
Flow Cytometry
905705
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, SLC12A5, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5
Mouse
IgG1
RUO
Primary
57468
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)