Learn More
KCC2/SLC12A5 Mouse anti-Human, Mouse, Rat, HRP, Clone: S1-12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259337H
Catalog No. NBP259337H
Add to cart
Description
Description
KCC2/SLC12A5 Monoclonal antibody specifically detects KCC2/SLC12A5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
KCC2/SLC12A5
S1-12
Western Blot, Immunohistochemistry
Purified
SLC12A5
Fusion protein amino acids 932-1043 corresponding to rat KCC2
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 140kDa.
Western Blot, Immunohistochemistry
HRP
PBS with No Preservative
Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5
Mouse
IgG2a
100 ul
Primary
57468
Human, Mouse, Rat