Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KChIP2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen KChIP2
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125642
View Documents
Novus Biologicals
NBP31037025UL
25 μg
Each for $106.19
Add to cart
 
NB125641
View Documents
Novus Biologicals
NBP310370100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

KChIP2 Polyclonal antibody specifically detects KChIP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

KChIP2
Western Blot, Immunohistochemistry
Purified
RUO
Human, Mouse
A-type potassium channel modulatory protein 2, cardiac voltage gated potassium channel modulatory subunit, Cardiac voltage-gated potassium channel modulatory subunit, DKFZp566L1246, KChIP2, KCHIP2Kv channel-interacting protein 2, Kv channel interacting protein 2, MGC17241, potassium channel interacting protein 2, Potassium channel-interacting protein 2
The immunogen is a synthetic peptide directed towards the N terminal region of human KChIP2 (NP_775285). Peptide sequence QLTDSVDDEFELSTVCHRPEGLEQLQEQTKFTRKELQVLYRGFKNPGALS
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Vision
PBS buffer, 2% sucrose
30819
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit