Learn More
KCNA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181031AP
Catalog No. 50-173-1987
Add to cart
Description
Description
KCNA10 is a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It is specifically regulated by cGMP and postulated to mediate the effects of substances that increase intracellular cGMP. This gene is intronless, and the gene is clustered with genes KCNA2 and KCNA3 on chromosome 1.Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It is specifically regulated by cGMP and postulated to mediate the effects of substances that increase intracellular cGMP. This gene is intronless, and the gene is clustered with genes KCNA2 and KCNA3 on chromosome 1.
Specifications
Specifications
KCNA10
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B2RQA1, Q16322
Kcn1, KCNA10, Kv1.8
Rabbit
IgG
150 μL
-20°C
Primary
242151, 295360, 3744
Immunohistochemistry (Paraffin)
Unconjugated
KCNA10
Liquid
KCNA10
KCNA10 Fusion Protein Ag12434
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat