Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNA10 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen KCNA10
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095933
View Documents
Novus Biologicals
NBP304425100UL
100 μg Each for $320.71
Add to cart
 
NB095934
View Documents
Novus Biologicals
NBP30442520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

KCNA10 Polyclonal antibody specifically detects KCNA10 in Human samples. It is validated for Western Blot
Specifications

Specifications

KCNA10
Unconjugated
Rabbit
Human
cyclic GMP gated potassium channel, Kcn1, Kv1.8, potassium voltage-gated channel subfamily A member 10, potassium voltage-gated channel, shaker-related subfamily, member 10, Voltage-gated potassium channel subunit Kv1.8
Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human KCNA10 (NP_005540.1).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
3744
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit