Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNAB1 Mouse anti-Human, Clone: OTI7C7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503968

Catalog No. 50-167-4741


Add to cart

Description

Description

KCNAB1 is a protein involved in potassium channels representing the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNAB1 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily.
Specifications

Specifications

KCNAB1
Monoclonal
0.69 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14722
AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7881
Immunohistochemistry (Paraffin), Western Blot
OTI7C7
Unconjugated
KCNAB1
Liquid
KCNAB1
Full length human recombit protein of human KCNAB1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit