Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnc2 Mouse anti-Mouse, Rat, PE/ATTO 594, Clone: S410-17, Abnova™

Mouse Monoclonal Antibody.

Manufacturer:  Abnova Corporation MAB19182

Catalog No. 89-338-687


Add to cart

Description

Description

Kcnc2 Monoclonal antibody specifically detects Kcnc2 in Mouse, Rat samples. It is validated for ICC, WB-Ti, IHC-P, IF.
Specifications

Specifications

Kcnc2
S410-17
potassium voltage gated channel, Shaw-related subfamily, member 2
Liquid
Mouse
IgG1
Store at -20°C
Monoclonal
Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
PE/ATTO 594
Kcnc2
KShIIIA, Kv3.2
Recombinant protein corresponding to amino acids 474-613 at C-terminus of rat Kcnc2
100 μg
Primary
246153
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit