Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnc2, Mouse anti-Rat, Clone: S410-17, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB16468

Catalog No. 89-999-425


Add to cart

Description

Description

Specifications

Specifications

Kcnc2
S410-17
Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
potassium voltage gated channel, Shaw-related subfamily, member 2
KShIIIA/Kv3.2
Mouse
IgG1
RUO
Antibody
Monoclonal
Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Kcnc2
Recombinant protein corresponding to amino acids 474-613 at C-terminus of rat Kcnc2.
100 μg
Store at -20°C.
Primary
246153
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit