Learn More
KCNC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 180921AP
Catalog No. 50-173-1980
Add to cart
Description
Description
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes. It generates atypical voltage-dependent transient current that may be important for neuronal excitability. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
KCNC4
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q03721, Q63734, Q8R1C0
HKSHIIIC, KCNC4, KSHIIIC, KV3.4
Rabbit
IgG
150 μL
-20°C
Primary
3749, 684516, 99738
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KCNC4
Liquid
KCNC4
KCNC4 Fusion Protein Ag12815
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat