Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen KCNE1
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094285
View Documents
Novus Biologicals
NBP304405100UL
100 μg
Each for $320.71
Add to cart
 
NB094286
View Documents
Novus Biologicals
NBP30440520UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

KCNE1 Polyclonal antibody specifically detects KCNE1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

KCNE1
Unconjugated
Rabbit
Human, Mouse, Rat
Delayed rectifier potassium channel subunit IsK, FLJ18426, FLJ38123, IKs producing slow voltage-gated potassium channel subunit beta Mink, ISK, Isk-related subfamily, member 1, JLNS2FLJ94103, LQT2/5, MGC33114, Minimal potassium channel, MinK, potassium voltage-gated channel, Isk-related family, member 1, voltage gated potassiun channel accessory subunit
Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human KCNE1 (NP_001121142.1).
Polyclonal
Affinity purified
Immunohistochemistry
Purified
RUO
PBS with 50% glycerol, pH7.3.
3753
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit