Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNE4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549024
Description
Predicted to react with mouse based on sequence homology.
Ancillary protein that assembles as a beta subunit with a voltage-gated potassium channel complex of pore-forming alpha subunits. Modulates the gating kinetics and enhances stability of the channel complex. May associate with KCNQ1/KVLTQ1 and inhibit potassium current.Specifications
KCNE4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8WWG9 | |
KCNE4 | |
KLH conjugated synthetic peptide between 58-86 amino acids from the Central region of human KCNE4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KCNE4 | |
Potassium voltage-gated channel subfamily E member 4, MinK-related peptide 3, Minimum potassium ion channel-related peptide 3, Potassium channel subunit beta MiRP3, KCNE4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23704 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction