Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550691

Catalog No. PA550691


Add to cart

Description

Description

The antibody detects endogenous levels of total KCNG3 protein.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member is a gamma subunit functioning as a modulatory molecule.
Specifications

Specifications

KCNG3
Polyclonal
Unconjugated
KCNG3
Liquid
KCNG3
Synthetic peptide corresponding to a region derived from internal residues of human potassium voltage-gated channel, subfamily G, member 3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
2.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8TAE7, P59053, Q8R523
KV6.3, KV10.1
Rabbit
IgG
100 μL
-20°C
Primary
170850, 171011, 225030
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit