Learn More
KCNH1 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS2424R
Catalog No. 50-198-4716
Add to cart
Description
Description
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. KCNH1 encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit of a voltage-gated non-inactivating delayed rectifier potassium channel. KCNH1 is activated at the onset of myoblast differentiation. KCNH1 is highly expressed in brain and in myoblasts. Overexpression of KCNH1 may confer a growth advantage to cancer cells and favor tumor cell proliferation. Alternative splicing of KCNH1 results in two transcript variants encoding distinct isoforms.
Specifications
Specifications
KCNH1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KCNH1
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
3756
KCNH1
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
O95259
Eag; EAG channel 1; EAG1; ether a go-go; Ether-a-go-go potassium channel 1; ether-a-go-go, Drosophila, homolog of; h-eag; hEAG1; KCNH1; Kv10.1; M-eag; potassium channel, voltage gated eag related subfamily H, member 1; Potassium voltage-gated channel subfamily H member 1; potassium voltage-gated channel, subfamily H (eag-related), member 1; r-eag; TMBTS; Voltage-gated potassium channel subunit Kv10.1; ZLS1
KLH conjugated synthetic peptide derived from human KCNH1.
Protein A
RUO
Antibody
Polyclonal
Human