Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNH6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104027
Description
Antibody detects endogenous levels of total KCNH6.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. KCNH6 encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit.
Specifications
KCNH6 | |
Polyclonal | |
Unconjugated | |
KCNH6 | |
Eag-related gene member 2; eag-related protein 2; Erg2; ERG-2; Ether-a-go-go-related gene potassium channel 2; ether-a-go-go-related protein 2; HERG2; hERG-2; KCNH6; Kv11.2; m-erg2; potassium channel, voltage gated eag related subfamily H, member 6; potassium voltage-gated channel subfamily H member 6; potassium voltage-gated channel, subfamily H (eag-related), member 6; voltage-gated potassium channel subunit Kv11.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
116745, 192775, 81033 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O54853, Q9H252 | |
KCNH6 | |
A synthesized peptide derived from human KCNH6(Accession Q9H252), corresponding to amino acid residues P884-E934. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction