Learn More
Aviva Systems Biology Corp KCNH7 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP35379_P050
This is a rabbit polyclonal antibody against KCNH7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNH7. Alternate names: ERG3, HERG3, Kv11.3, MGC45986ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-55719