Learn More
KCNIP1 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00030820T01
Catalog No. 89-002-172
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
BC050375
6.8
Transient overexpression cell Lysate was tested with Anti-KCNIP1 antibody (H00030820-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
KCNIP1 293T Cell Transient Overexpression Lysate(Denatured)
23.87
pCMV-KCNIP1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.